Learn More
Taylor Wharton™ CXR100 Cryogenic Dry Shipper
Product Code. 11748419
Change view
Click to view available options
Capacity (Metric):
3.7 L Liquid Nitrogen
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11748419 3.7 L Liquid Nitrogen Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11748419 Supplier Taylor Wharton™ Supplier No. CXR100
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
Estimated Shipment: 12-04-2027
Designed to safely transport a wide variety of materials at cryogenic temperatures. Taylor Wharton™ CXR100 Cryogenic Dry Shipper features a unique absorbent material that prevents a liquid spill if the unit is tipped over.
  • Advanced Concept Absorbent enables faster charging
  • Complies with IATA regulation
  • New rugged construction includes dependable, stronger, magniformed necktube design, ribbed high strength aluminum body, and more durable paint system
  • Superior vacuum performance with super insulation provides maximum holding times
  • Optional storage accessories include a five shelf rack system for the CX500
  • Lockable lids
  • Warranty: 2 years on vacuum, 1 year on electronic
Specifications
Capacity (Metric) 3.7 L Liquid Nitrogen
Height (Metric) Exterior 493 mm
Width (Metric) Exterior 234 mm
Vial Capacity 102 Vials
Weight (Metric) 5.3/8.3 kg (Empty/Full)