Learn More
Taylor Wharton DRY SHIPPER CXR500 7,7L
Brand:  Taylor Wharton CXR5000
710.93 EUR valid until 2026-01-16
BEST PRICE on Promo! Use promo code "24103" to get your promotional price.
DRY SHIPPER CXR500 7,7L
Product Code. 11778419
  • 3485.00€ / Each
Outlet Corner
Use promo code 24103