Learn More
Welch Ilmvac™ Diaphragm Vacuum Pump Service and Maintenance Kit
Shop All Welch Ilmvac Products
Click to view available options
For Use With:
MP/MPC 101 Z, 201 E
MP/MPC 301 Z, 601 E, 301 Z
MP/MPC 601 T, 901 Z, 1201 E, MP 301V
MP/MPR 060 E, 030 Z, Biovac 060, MP 101V
Unit Size:
Each
Description
- Available for a range of diaphragm vacuum pump models
- Provided individually
Specifications
Specifications
| For Use With | MP/MPR 060 E, 030 Z, Biovac 060, MP 101V |
| Includes | Diaphragm, Valves, and O-rings |
| Quantity | 1 Ea. |
| Type | Service Kit |