Learn More
Equip Labo™ Dry Bare Benches
Brand:  Equip Labo™ 8096
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12568335
  • 1275.00€ / Each
Estimated Shipment: 20-06-2024
Specifications
750 x 1800 x 900 mm
Dry Bare Bench
750 mm
180 cm
12 Months
900 mm
130 kg
150 mm