Learn More
Equip Labo™ Dry Bare Benches
615.00€ - 1275.00€
Specifications
Load Bearing Capacity (Metric) | 130 kg |
---|---|
Warranty | 12 Months |
Product Type | Dry Bare Bench |
Product Code | Brand | Depth (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
12556506
|
Equip Labo™
8000 |
600 mm | 600 mm |
615.00€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
12557806
|
Equip Labo™
8010 |
600 mm | 900 mm |
715.00€
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
12587696
|
Equip Labo™
8020 |
600 mm | 120 cm |
765.00€
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
12566506
|
Equip Labo™
8030 |
600 mm | 150 cm |
855.00€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
12558335
|
Equip Labo™
8070 |
750 mm | 120 cm |
835.00€
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
12507806
|
Equip Labo™
8080 |
750 mm | 150 cm |
940.00€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
12568335
|
Equip Labo™
8096 |
750 mm | 180 cm |
1275.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
Specifications
130 kg | |
12 Months | |
Dry Bare Bench |