Learn More
Equip Labo™ Dry Bare Benches
Brand:  Equip Labo™ 8030
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12566506
  • 855.00€ / Each
Estimated Shipment: 27-06-2024
Specifications
600 x 1500 x 900 mm
Dry Bare Bench
600 mm
150 cm
12 Months
900 mm
130 kg
150 mm