Learn More
Equip Labo™ Dry Bare Benches
Brand:  Equip Labo™ 8070
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12558335
  • 835.00€ / Each
Estimated Shipment: 19-06-2024
Specifications
750 x 1200 x 900 mm
Dry Bare Bench
750 mm
120 cm
12 Months
900 mm
130 kg
150 mm