Learn More
Equip Labo™ Dry Bare Benches
Brand:  Equip Labo™ 8010
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12557806
  • 715.00€ / Each
Estimated Shipment: 31-05-2024
Specifications
12 Months
600 mm
900 mm
Dry Bare Bench
130 kg