Learn More
Equip Labo™ Dry Bare Benches
Brand:  Equip Labo™ 8000
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12556506
  • 615.00€ / Each
Estimated Shipment: 28-06-2024
Specifications
12 Months
600 mm
600 mm
Dry Bare Bench
130 kg