Learn More
Equip Labo™ Dry Bare Benches
Brand:  Equip Labo™ 8080
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12507806
  • 940.00€ / Each
Estimated Shipment: 19-06-2024
Specifications
Glass Emalite Worktop
12 Months
900 mm
130 kg
150 mm
750 x 1500 x 900 mm
Dry Bare Bench
750 mm
150 cm