Learn More
asecos™ Drum Rack With Sump Pallet Steel
Product Code. 17866366 Shop All Asecos Products
Change view
Click to view available options
Weight (Metric):
66 kg
73 kg
118 kg
139 kg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
17866366 118 kg Each
17856416 66 kg Each
17836406 73 kg Each
17806426 139 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17866366 Supplier asecos™ Supplier No. 13089
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Drum Rack With Sump Pallet Steel provides safe storage and filling storage of substances hazardous to water of all hazard classes.
  • For the storage of oil, paint and lacquer.
  • Safe dispensing due to overhanging collecting sump.
Specifications
Capacity (English) 54.2 gal.
Capacity (Metric) 205 L
Material Steel
Type Drum Rack
Length (English) 52.36 in.
Length (Metric) 1330.00 mm
Width (English) 52.36 in.
Width (Metric) 1330.00 mm
Height (English) 61.81 in.
Height (Metric) 1570.00 mm
Depth (English) 32.09 in.
Depth (Metric) 815.00 mm
Weight (English) 260.14 lb.
Weight (Metric) 118 kg