Learn More
Buerkle™ Drum Pump for Mineral Oils
Shop All Bürkle Products
Click to view available options
Flow Rate:
85 L/min.
Unit Size:
Each
Description
- Motor 230 V
- Pump tube made of aluminum
- Hose connection Alu 1 in.
- 2 m PVC fabric hose 1 in.
- Hose clips 1 in.
- Alu pump nozzle, PP barrel screw joint
- Medium-contacting parts made of aluminum and AISI 316 Ti (1.4571)
- Delivery pump height 19 mWS, Pump capacity 85 LPM
Specifications
Specifications
| Voltage | 230 V |
| Flow Rate | 85 L/min. |
| Includes | Hose connection Alu 1 in.; 2 m PVC fabric hose 1 in.; Hose clips 1 in.; Alu pump nozzle, PP barrel screw joint |
| For Use With | Mineral Oils |
| Pump Tube Length | 100 cm (Tube dia. 41 mm) |
| Material | Aluminum, AISI 316 Ti (1.4571) |