Learn More
Oakton™ Economy Hygrothermograph
Product Code. 11708639 Shop All Cole-Parmer Products
Change view
Click to view available options
Product Type:
Red Felt-tip Pen
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11708639 Red Felt-tip Pen Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11708639 Supplier Oakton™ Supplier No. 0836880
Estimated Shipment: 11-12-2026
Use these replacement pens with Oakton™ Economy Hygrothermographs.
  • Red felt tip pens
  • Sold in packs of six
  • For Oakton™ Economy Hygrothermograph
Specifications
Includes 55 Sheets of seven-day, chart paper and one AA battery
For Use With (Application) Measuring and recording both humidity and temperature
Product Type Red Felt-tip Pen