Learn More
Techne™ Dri-Block™ Flat Bottom Plate Insert
Description
- For use with Techne™ dri-block heaters
- Manufactured from anodised aluminum
- Blocks can be used in any combination
Specifications
Specifications
| Description | Flat-Bottom |
| For Use With (Equipment) | Techne Dri-Block Heaters |
| Material | Anodized Aluminum |
| Dimensions (D x W x H) Interior | 95 x 150 x 50 mm |