missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DREADD agonist 21 dihydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
DREADD agonist 21 dihydrochloride is a water soluble version of DREADD agonist 21 (Cat. No. 5548). Potent muscarinic DREADD agonist.
Specifications
Specifications
| Chemical Name or Material | 11-(1-Piperazinyl)-5 H-dibenzo[b,e][1,4 ]diazepine dihydrochloride |
| CAS | 2250025-92-2 |
| Quantity | 10 mg |
| Target | DREADD Activators |
| Molecular Formula | C17H18N40.2 HCl |
| Formula Weight | 351.27 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction