Learn More
Zeiss™ Drawing Intermediate Tube
Product Code. 11845133
Click to view available options
Type:
Drawing Intermediate Tube S
Unit Size:
Each
Product Code. 11845133
Brand: Zeiss™ 435111
This item requires a price update prior to being ordered.
Request a Price for this item here.
Switchable for stereoscopic viewing of specimen or for binocular 2D viewing of specimen and drawing
Compatible Microscopes:
  • SteREO Discovery.V12
  • SteREO Discovery.V20
  • SteREO Discovery.V8
  • SteREO Lumar.V12
Specifications
Type Drawing Intermediate Tube S
For Use With (Application) Stereoscopic viewing or binocular 2D viewing and drawing
Quantity 1 Ea.
Size Small