Learn More
Olympus™ Drawing Attachment Tube, SZX
Product Code. 12542357
Click to view available options
For Use With (Equipment):
SZX microscopes
Unit Size:
Each
Product Code. 12542357
Brand: Olympus™ K10017133
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Create drawings from microscope views using this attachment arm. Olympus™ Drawing Attachment Tube, SZX mounts between the viewing head and the objectives and projects an image to a sheet for drawing/tracing.
  • Light intensity ratio: 50% observation, 50% drawing.
  • Focusing range: 200mm (working distances 250 to 450mm).
  • Intermediate magnification: 1X
  • Drawing magnification: 6.3 x total magnification on zooming knob/10
    (with the standard base, 1X objective and 10X eyepiece)
  • Drawing size: 138mm dia.
    (with the standard base, 1X objective and 10X eyepiece).
Specifications
For Use With (Equipment) SZX microscopes