Learn More
Thermo Scientific™ Drawer Dividers
Tienda Thermo Scientific Productos
Click to view available options
For Use With (Equipment):
4.9 and 11.5 cu. ft. Thermo Scientific Revco and Forma blood bank refrigerators and plasma freezers.
Tamaño de la unidad:
Each
Descripción
Maximize storage inside blood bank refrigerators and plasma freezers with these optional Thermo Scientific™ Drawer Dividers.
Stainless-steel drawer dividers help manage inventory of bags and boxes:
- Four column dividers for all models except 29.2 cu. ft. (826L) models
- Five column dividers for 29.2 cu. ft. (826L) models
Especificaciones
Especificaciones
| For Use With (Equipment) | 4.9 and 11.5 cu. ft. Thermo Scientific Revco and Forma blood bank refrigerators and plasma freezers. |
| Holds | Bags |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido