missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ DRAQ9™

Product Code. 18060914 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μL
1000 μL
Unit Size:
1000µL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18060914 200 μL 200µL
18247298 1000 μL 1000µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18060914 Supplier Novus Biologicals™ Supplier No. NBP281128200UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Content And Storage -20°C
Quantity 200 μL
For Use With (Application) Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.