Specifications
| Content And Storage | Store at 4°C in the dark. Do not freeze. |
| Quantity | 50 μL |
| For Use With (Application) | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy |