Learn More
Dräger™ Short-Term Detector Tubes: Ammonia
Product Code. 10695431
Change view
Click to view available options
Measurement Range:
0.25 to 3
0.5 to 10%
2 to 30
5 to 100
5 to 700
Tube Type:
Ammonia 0.25/a
Ammonia 0.5%/a
Ammonia 2/a
Ammonia 5/a
Ammonia 5/b
Humidity Range:
3 to 12mg/L
Less than20mg/L
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Tube Type Humidity Range Unit Size
10695431 5 to 700 Ammonia 5/a Less than20mg/L Pack of 10
10060042 0.25 to 3 Ammonia 0.25/a Less than20mg/L Pack of 10
10110971 5 to 100 Ammonia 5/b Less than20mg/L Pack of 10
10299941 2 to 30 Ammonia 2/a Less than20mg/L Pack of 10
15220578 2 to 30 Ammonia 2/a Less than20mg/L Each
15132323 0.5 to 10% Ammonia 0.5%/a 3 to 12mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10695431 Supplier Dräger™ Supplier No. CH20501
  • ONLINE EXCLUSIVE PRICE
    184.25€
    143.00€ / Pack of 10
    Save 41.25€
    22% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Description Detector Tubes
Components Measured Ammonia
For Use With (Equipment) Draeger Accuro Pump
Type Gas Detection Tube
Humidity Range Less than20mg/L
Measurement Range 5 to 700
No. of Strokes 10
Quantity 10
Response Time 1 min.
Temperature Range (English) 50°F to 122°F
Temperature Range (Metric) 10° to 50°C
Tube Type Ammonia 5/a