Learn More
Hirschmann™ Automatic Burettes, Acc. to Dr. Schilling
Product Code. 10569222
Click to view available options
Capacity (Metric):
5 mL
10 mL
15 mL
25 mL
50 mL
Unit Size:
Each
Product Code. 10569222
Brand: Hirschmann™ 3600164
Excellent for automatically dispensing measured amounts of chemical solutions in the laboratory. Hirschmann Instruments™ Automatic Burettes, Acc. to Dr. Schilling are available in a range of capacities to fit your needs.
  • With Schellbach stripe
  • Blue graduation
  • Fast flow burette
Specifications
Capacity (Metric) 15 mL
Material Glass
Graduations 1 mL
Includes LDPE Bottle
Tolerance 0.045
Type Titration Apparatus
Certifications/Compliance Batch Certified, Individual Certificate, DKD Certificate