Learn More
Draeger Safety DrögerSensor™ XXS CO LC
Product Code. 15600899
Change view
Click to view available options
Measurement Range:
0 to 2000 ppm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
15600899 0 to 2000 ppm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15600899 Supplier Draeger Safety Supplier No. 6813210
  • ONLINE EXCLUSIVE PRICE
    543.15€
    425.73€ / Pack of 1
    Save 117.42€
    21% Off
Estimated Shipment: 15-12-2026
Draeger Safety DrögerSensor™ XXS CO LC detects CO at 1 ppm resolution for use with X-am™ 5600 series multi-gas monitors.
  • 0 - 2000 ppm gas detector sensor
  • Detects CO at 1 ppm resolution
  • For use with X-am™ 5600 series multi-gas monitors
Specifications
Certifications/Compliance CSA Certified, MED Approved
Components Measured Carbon Monoxide
For Use With (Equipment) X-am 5600 Series Multigas Monitors
Measurement Range 0 to 2000 ppm
Resolution 1 ppm
Temperature Range (Metric) -20°C to +50°C