Learn More
Dräger™ XS Sensors for MiniWarn™ Multi-Gas Monitors
Product Code. 10030032
Change view
Click to view available options
Measurement Range:
0 to 200
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10030032 0 to 200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10030032 Supplier Dräger™ Supplier No. 6809115
  • ONLINE EXCLUSIVE PRICE
    1506.98€
    1186.94€ / Pack of 1
    Save 320.04€
    21% Off
Estimated Shipment: 22-12-2026
For detecting or monitoring acetaldehyde gas
  • Range: 0 to 200 ppm with 1 ppm resolution
Specifications
Components Measured Organic Vapor
Humidity Range 10 to 90%
Measurement Range 0 to 200
Pressure Range 700 to 1300mbar
Resolution 0.5
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Warranty 1 Year