Learn More
Dräger™ Short-Term Detector Tubes - Ozone
For making spot measurements for various gases and vapors
237.00€ - 254.00€
Product Code | Brand | Components Measured | Measurement Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Components Measured | Measurement Range | Price | Quantity & Availability | ||||
10120921
|
Dräger™
6733181 |
Ozone 0.05/b | 0.05 to 0.7ppm |
254.00€
Pack of 10 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
10355142
|
Dräger™
CH21001 |
Ozone 10/a | 20 to 300ppm |
237.00€
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
Description
- Glass vial
- Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
1Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
2Safety Equipment Institute (SEI) certified.