Learn More
Dräger™ Short-Term Detector Tubes - Nitrogen Fumes
Click to view available options
Components Measured:
Nitrous Fumes 100/c
Nitrous Fumes 2/a
Nitrous Fumes 20/a
Measurement Range:
100 to 5000ppm
2 to 100ppm
20 to 500ppm
Unit Size:
Pack of 10
Description
- Glass vial
- Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
Specifications
| Components Measured | Nitrous Fumes 100/c |
| Humidity Range | 30mg/L |
| Measurement Range | 100 to 5000ppm |
| No. of Strokes | 5/1+4 |
| Quantity | 10 |
| Response Time | 1.5 min./18 sec. |
| Temperature Range (English) | 50°F to 86°F |
| Temperature Range (Metric) | 10°C to 30°C |