Learn More
Dräger™ Short-Term Detector Tubes: Formaldehyde Activation Tube
Product Code. 10396292
Change view
Click to view available options
Measurement Range:
0.04
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
10396292 0.04 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10396292 Supplier Dräger™ Supplier No. 8101141
  • ONLINE EXCLUSIVE PRICE
    191.70€
    148.95€ / Pack of 10
    Save 42.75€
    22% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Formaldehyde
Measurement Range 0.04
Quantity 10
Tube Type Formaldehyde activation tube (used w/Formaldehyde 0.2/a)