Learn More
Dräger™ Flow Check Air Flow Indicator Kit
Product Code. 10013853
Change view
Click to view available options
Type:
Air Indicator Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10013853 Air Indicator Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10013853 Supplier Dräger™ Supplier No. 6400761
  • ONLINE EXCLUSIVE PRICE
    1688.03€
    1369.41€ / Pack of 1
    Save 318.62€
    18% Off
Estimated Shipment: 10-12-2026
Includes
Three ampules, a nozzle, and rechargeable NiCd batteries that offer about 20 minutes of operation per charge
Generates a plume of smoke, making the direction and dispersion of air currents visible, as well as their relative speed
Ideal for any applications where ventilation controls the health and safety of workers, such as paint spray booths and fume hoods. Also useful tool for air balancing, adjusting ventilation louvers in offices and institutions, and performing indoor air quality studies. Acts as a leak detector by using smoke to check airflow outside airtight doors, detecting leaks into cleanrooms or containment areas.
  • Lightweight, ergonomically designed unit offers easy, one-handed operation
  • Generates environmentally safe smoke in puffs or in a continuous cloud
  • Each smoke-generating ampule provides about five minutes of smoke
Order Info
Measures approximately 11 3/4 x 7 x 2 3/4 in. (300 x 200 x 70mm) and weighs 171/2 oz. (500g). Chargers and replacement ampules are available separately. For charger, see Cat. No. 19-130-1060.
Specifications
Type Air Indicator Kit