Learn More
Dräger™ Connecting cradle for Pac™ 1000, 3500, 5500, and 7000 Series Gas Detectors
Product Code. 15120629
Change view
Click to view available options
For Use With (Equipment):
Pac 1000, 3500, 5500, and 7000 Series Gas Detectors
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15120629 Pac 1000, 3500, 5500, and 7000 Series Gas Detectors Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15120629 Supplier Dräger™ Supplier No. 8318587
  • ONLINE EXCLUSIVE PRICE
    429.20€
    348.30€ / Pack of 1
    Save 80.90€
    18% Off
Estimated Shipment: 19-01-2027
Includes
USB cable
Accessory for Pac series gas detectors
  • Allows connection of any Pac detector to PC
Specifications
Dimensions (L x W x H) 10 x 8 x 1 in. (304.8 x 20.32 x 2.5cm)
For Use With (Equipment) Pac 1000, 3500, 5500, and 7000 Series Gas Detectors
Type Connecting Cradle
Height (English) 1 in.
Height (Metric) 2.5 cm
Length (English) 10 ft.
Length (Metric) 304.8 cm
Width (English) 8 in.
Width (Metric) 20.32 cm