Learn More
Dräger™ CMS Chips: Ammonia
Product Code. 10436301
Click to view available options
Measurement Range:
0.2 to 5
10 to 150
2 to 50
Response Time:
100 to 600 sec.
15 to 140 sec.
15 to 50 sec.
Humidity Range:
1 to 30mg/L
1 to 40mg/L
Unit Size:
Each
Product Code. 10436301
Brand: Dräger™ 6406020
Suitable for 10 measurements of one gas
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Ammonia CMS Chip
Humidity Range 1 to 40mg/L
Measurement Range 10 to 150
Response Time 15 to 50 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C