Learn More
Dräger™ CMS Chips: Hydrogen Sulfide
Product Code. 12304629
Click to view available options
Measurement Range:
0.2 to 5
100 to 2500
2 to 50
20 to 500
Response Time:
20 to 200 sec.
30 to 240 sec.
40 to 450 sec.
40 to 500 sec.
Humidity Range:
1 to 30mg/L
1 to 40mg/L
Unit Size:
Each
Product Code. 12304629
Brand: Dräger™ 6406220
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Suitable for 10 measurements of one gas.
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Hydrogen Sulphide CMS Chip
Humidity Range 1 to 40mg/L
Measurement Range 100 to 2500
Response Time 40 to 500 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C