Learn More
Dräger™ Alkaline Battery Holder for X-am™ 1000/2000/5000 Gas Monitors
Product Code. 15112697
Change view
Click to view available options
For Use With (Equipment):
X-am 1000/2000/5000 Gas Detectors
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15112697 X-am 1000/2000/5000 Gas Detectors Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15112697 Supplier Dräger™ Supplier No. 8318703
  • ONLINE EXCLUSIVE PRICE
    85.20€
    70.00€ / Pack of 1
    Save 15.20€
    17% Off
 Restricted Item
Estimated Shipment: 19-01-2027
Accessory for X-am 1000/2000/5000 Gas Detectors.
Accessory for X-am 1000/2000/5000 Gas Monitors.
Specifications
Dimensions (L x W x H) 3 x 1.5 x 0.5 in. (7.6 x 3.8 x 1.2cm)
For Use With (Equipment) X-am 1000/2000/5000 Gas Detectors
Type Alkaline Battery Holder
Height (English) 0.5 in.
Height (Metric) 1.2 cm
Length (English) 3 in.
Length (Metric) 7.6 cm
Voltage 110V
Width (English) 1.5 in.
Width (Metric) 3.8 cm