missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DPTIP

Description
DPTIP is a potent and selective sphingomyelin phosphodiesterase (N-SMase2) inhibitor (IC50 = 30 nM). Exhibits no inhibition of related enzymes alkaline phosphatase, phosphomonoesterase or acid sphingomyelinase. Inhibits extracellular vesicle (EV) release in primary astrocyte cultures and in vivo. Attenuates IL-1β-induced astrocyte-derived EV release in a mouse model of brain injury and suppresses infiltration of immune cells into the brain. Metabolically stable and brain penetrant.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C21H18N2O3S |
| Formula Weight | 378.45 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?