missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dowex Marathon C, H+ form, Thermo Scientific™
Click to view available options
Quantity:
250 g
1000 g
2500 g
Unit Size:
1000g
2500g
250g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is designed for demineralization using HCl or H2 SO4 as regenerate in co-current or counter-current operation.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
It is designed for demineralization using HCl or H2 SO4 as regenerate in co-current or counter-current operation.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Chemical Name or Material | Dowex Marathon C |
| Name Note | H+ form |
| CAS | 150604-77-6 |
| MDL Number | MFCD00212649 |
| Density | 1.2 |
| Quantity | 1000 g |
| Solubility Information | Insoluble in water. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction