Learn More
Brand™ Counting Chambers, Fuchs-Rosenthal pattern, BLAUBRAND™, Double ruling without clips
Product Code. 15623997 Shop All Brand Products
Change view
Click to view available options
Depth (Metric):
0.2 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
15623997 0.2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15623997 Supplier Brand™ Supplier No. 719805
  • ONLINE EXCLUSIVE PRICE
    63.00€
    56.18€ / Pack of 1
    Save 6.82€
    10% Off
 Restricted Item
In Stock
BLAUBRAND™ quality. Outstanding measurement precision. 100% certified quality.
  • The ruling differs from the customary systems for blood cell counts by its large area of 16 mm2
  • The ruling shows 16 large squares of 1 mm2.
  • Each large square is subdivided into 16 mini squares with 0.25 mm sides and an area of 0.0625 mm2
  • This chamber is frequently used for counting cells in the cerebro-spinal fluid.
TRUSTED_SUSTAINABILITY
Specifications
Type Counting Chamber Fuchs-Rosenthal
Culture Area 16 mm2
CE Approved CE-marked according to IVD 98/79 EG.
Depth (Metric) 0.2 mm