Learn More
Wesbart™ Double Platform Test Tube Rack
Product Code. 15723368
Click to view available options
Positions:
25
40
50
54
100
145
Holds:
Tubes (14 mm)
Tubes (15 mL), Tubes (17 mm)
Tubes (18 mm)
Tubes (20 mm)
Tubes (30 mm)
Tubes (5 mL)
Unit Size:
Each
Product Code. 15723368
Brand: Wesbart™ DP3025
Only null left
Add to basket
Add to basket
Test Tube racks are an effective way of storing Falcon/ Universal tubes, Bijous, cryogenic vials or any size glass bottles.
  • Racks are available in aluminum and stainless steel with the option of a hand grip
  • Test Tube racks can be designed to fit in horizontal cold store or be free standing
  • Different size test tubes racks are available to suit your requirements
  • Labels can be fitted to racks so they can be easily identified
Specifications
Product Type Double Platform Test Tube Rack
Holds Tubes (30 mm)
Material Aluminum
Positions 25
Width (Metric) 182 mm
Height (Metric) 70 mm
Depth (Metric) 182 mm
Diameter (Metric) Opening 30 mm