Learn More
Metro™ Double-Deep qwikTRAK™ Mobile Unit Kits
Description
- For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax i™ shelving systems
- Kit includes eight 74 in. (188cm) high -UP posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Order Info
Order one kit per double-deep system. Shelves are sold separately.
Specifications
Specifications
| Product Type | Mobile Unit Kit |
| Finish | Chrome |
| For Use With (Equipment) | Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ shelving system |
| Includes | Eight 74 in. high up posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware |
| Width (English) Shelf | 21 in. |
| Width (English) | 21 in. |
| Width (Metric) Shelf | 53 cm |
| Width (Metric) | 53 cm |
| Material | Steel |
Product Title
¿Detecta una oportunidad de mejora?