Learn More
Micronova™ Double-Coated Liner Tape
Product Code. p-200109620 Shop All Micronova Products
Click to view available options
Adhesive Type:
Acrylic
Rubber
Unit Size:
36 rolls
Product Code. 18811411
Brand: Micronova™ DCL1TRH
Only null left
Add to basket
Add to basket
Double-Coated Liner Tape is a premium, high performance, double-sided, polyester film tape for permanent and temporary attachment in cleanrooms.
  • Product Type: Liner Tape
  • Material: Polyester
  • Format: Double-Sided
  • Width: 2.5 cm (1 in.)
  • Length: 33 m (36 yd.)
  • For Use With (Application): Cleanroom Contamination Control
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 Certified Facility
Adhesive Type Acrylic
Color Transparent
Format Tape
Material Polyester
Product Type Tape
Sterility Non-sterile
For Use With (Application) Permanent and temporary attachment
Latex Free Yes
Quantity 36 Rolls/Cs. (Individually bagged)
Length (English) 36 yd.
Length (Metric) 33 m
Width (English) 1 in.
Width (Metric) 2.5 cm