Learn More
Dotriacontane, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 544-85-4 |
| Molecular Formula | C32H66 |
| MDL Number | MFCD00009411 |
| Quantity | 100 g |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC |
| Molecular Weight (g/mol) | 450.87 |
| Percent Purity | 98% |
| Chemical Name or Material | Dotriacontane |