missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dothiepin-d6 HCl (N,N-dimethyl-d6) (cis/trans mixture), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (EZ)-Dosulepin-D6 (N,N-dimethyl-D6) Hydrochloride |
| CAS | 1276545-35-7 |
| Molecular Formula | C19 D6 H15 N S . Cl H |
| Quantity | 0.005 g |
| Synonym | 3-Dibenzo[b,e]thiepin-11(6 H)-ylidene-N,N-di(methyl-d3)-1-propanamine hydrochloride, Dothiepin-d6 hydrochloride, Dosulepin-d6 hydrochloride, Dothiepin-d6 (N,N-dimethyl-d6) HCl (cis/trans mixture), (EZ)-Dosulepin-D6 (N,N-dimethyl-D6) Hydrochloride |
| InChI Formula | InChI=1 S/C19H21NS.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12 H,7,13-14H2,1-2H3;1 H/i1D3,2D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(CCC=C1c2ccccc2CSc3ccccc13)C([2 H])([2 H])[2 H] |
| IUPAC Name | 3-(6 H-benzo[c][1 ]benzothiepin-11-ylidene)-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 337.9397 |
| Formula Weight | 337.1538 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction