Learn More
Socorex™ Dosys Stainless Steel Dispensing Cannula
Product Code. 10282503
Change view
Click to view available options
Diameter (Metric) Outer:
0.8 mm
1.2 mm
Unit Size:
Each
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
10282503 1.2 mm Each
10039502 0.8 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10282503 Supplier Socorex™ Supplier No. 370.1250
Estimated Shipment: 28-12-2026
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Diameter (Metric) Outer 1.2 mm
Product Type Dispensing Cannula
Length (Metric) Needle 50 mm