Learn More
Socorex™ Dosys™ Premium Two-ring Automatic Syringe
Product Code. 10111542
Click to view available options
Volume (Metric):
0.1 to 1 mL
0.3 to 2 mL
0.5 to 5 mL
Unit Size:
Each
Product Code. 10111542
Brand: Socorex™ 164.0505
Dose with excellent reproducibility using a corrosion-resistant two-ring handle automatic syringe. Socorex™ Dosys™ Premium Two-ring Automatic Syringe fits either hand comfortably and requires limited effort for efficient work even during long dispensing sessions.
  • Two-ring handle.
  • Self-refilling automatic syringe.
  • Detachable Luer lock valve system.
  • High chemical resistance.
  • Corrosion resistant.
  • Optimal hand balance and ergonomics.
  • Fits either hand.
  • Requires limited effort.
  • Fast and reliable volume setting.
  • High reproducibility due to safety locking nut on a micrometric screw.
  • High-precision stainless steel plunger.
  • Ground glass barrel.
  • Smooth plunger travel and tightness without O-ring.
  • Easy disassembly for efficient cleaning and parts replacement if necessary.
  • Autoclavable (except for protective PVC sleeve).
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Includes Syringe, 1M silicone feed tube, sinker, PVC protection sleeve, aspiration, vent and dosing cannulas, set of spares for valve, operating instructions.
Product Type Automatic Syringe, Two-ring
Graduations 0.5 mL
Model Dosys™ Premium 164
Quantity 1/ea.
Precision <0.5% to <0.2%
Volume (Metric) 0.5 to 5 mL
Connection Type Luer Lock
Graduated Yes