Learn More
Hanna Instruments™ Dosing Pump
Product Code. 10458822
Click to view available options
Flow Rate:
1.5 L/hr.
10 L/hr.
20 L/hr.
5 L/hr.
7 L/hr.
Unit Size:
Each
Product Code. 10458822
Brand: Hanna Instruments™ BL202
Specifications
Overall Height (Metric) 150 mm
Overall Length (Metric) 180 mm
Temperature (Metric) Max 45°C
Flow Rate 20 L/hr.
Maximum Pressure 0.5 bar
Weight (Metric) 4 kg