Learn More
Doseheads for Sensititre AIM™ for plate inoculation
Product Code. 15456574 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15456574 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15456574 Supplier Thermo Scientific Supplier No. YE3010
  • ONLINE EXCLUSIVE PRICE
    334.00€
    220.37€ / Pack of 100
    Save 113.63€
    34% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-11-2026
Dose most 96-well microtitre plates, eliminating skipped wells and repeat tests, with the Thermo Scientific™ Doseheads for Sensititre AIM™ for plate inoculation.
Quickly and accurately dose most 96-well microtitre plates, eliminating both skipped wells and costly repeat tests, with the Thermo Scientific™ Doseheads for Sensititre AIM™ for plate inoculation.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description Doseheads for Sensititre for plate inoculation
Quantity 100/Pk.