Description
- Provides security for incubators and chambers
- Lockable door handle
Specifications
Specifications
| Description | Door lock - lockable door handle |
| For Use With (Equipment) | BINDER Chambers Series VD, VDL |
| Material | Plastic |
| Type | Door Handle Lock |