Learn More
Edmund Buhler Door Cabinet With Drawer
Click to view available options
Color:
White
Unit Size:
Each
Description
- Door cabinet with a drawer
- 4 cabinets 1 drawer + 2 doors
- Laboratory floor cabinets
- Width 84 cm
Specifications
Specifications
| Product Type | Door/Drawer Cabinet |
| Color | White |
| Cabinet Type | Door and Drawer Cabinet |
| Depth (Metric) | 45 cm |
| Dimensions (D x W x H) | 45 x 84 x 72 cm |
| Door Type | 2 Door |
| Height (Metric) | 72 cm |
| No. of Doors | 2 |
| No. of Shelves | 1 |
| Warranty | 12 Months |
| Show More |