missing translation for 'onlineSavingsMsg'
Learn More

Dofetilide-d4, TRC

Product Code. 30376874
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376874 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30376874 Supplier Toronto Research Chemicals Supplier No. TRCD5257021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dofetilide-d4
CAS 1189700-56-8
Molecular Formula C19H23D4N3O5S2
Quantity 1 mg
Synonym N-[4-[2-[Methyl[2-[4-[(methylsulfonyl)amino]phenoxy]ethyl]amino]ethyl]phenyl]-methanesulfonamide-d4, UK-68798-d4, Tikosyn-d4
InChI Formula InChI=1S/C19H27N3O5S2/c1-22(13-12-16-4-6-17(7-5-16)20-28(2,23)24)14-15-27-19-10-8-18(9-11-19)21-29(3,25)26/h4-11,20-21H,12-15H2,1-3H3/i14D2,15D2
SMILES CN(CCC1=CC=C(NS(=O)(C)=O)C=C1)C([2H])([2H])C([2H])([2H])OC2=CC=C(NS(C)(=O)=O)C=C2
Molecular Weight (g/mol) 445.59
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.