Learn More
Dodecanoic-2-13C Acid, CDN
Product Code. 30309643
Change view
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309643 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30309643 Supplier CDN Supplier No. CDNC31560.5G
 Restricted Item
Estimated Shipment: 09-11-2026
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dodecanoic-2-13 C Acid
CAS 287100-78-1
Molecular Formula 13 C C11 H24 O2
Quantity 0.5 g
Synonym Dodecanoic-2-13 C acid (9 CI), [2-13 C]Lauric acid, Dodecanoic acid-2-13 C
InChI Formula InChI=1 S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i11+1
SMILES CCCCCCCCCC[13 CH2]C(=O)O
IUPAC Name (213 C)dodecanoic acid
Molecular Weight (g/mol) 201.3104
Formula Weight 201.181 g/mol
Percent Purity 99 atom % 13 C, min 98% Chemical Purity