Learn More
Dodecanoic-12,12,12-d3 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Lauric Acid-d3 |
| CAS | 79050-22-9 |
| Molecular Formula | C12 D3 H21 O2 |
| Quantity | 1 g |
| Synonym | Dodecanoic-12,12,12-d3 Acid, Lauric-12,12,12-d3 Acid, Dodecanoic-12,12,12-d3 acid (9 CI), Lauric-12,12,12-d3 acid (6 CI), [12,12,12-2H3]Dodecanoic acid, Dodecanoic acid-12,12,12-d3, Lauric acid-12,12,12-d3 |
| InChI Formula | InChI=1 S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCCCCC(=O)O |
| IUPAC Name | 12,12,12-trideuteriododecanoic acid |
| Molecular Weight (g/mol) | 203.34 |
| Formula Weight | 203.1965 g/mol |
| Show More |