missing translation for 'onlineSavingsMsg'
Learn More

Dodecamethylcyclohexasiloxane, TRC

Product Code. 30368990
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368990 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368990 Supplier Toronto Research Chemicals Supplier No. TRCD494520250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dodecamethylcyclohexasiloxane
CAS 540-97-6
Purity Grade Notes GC
Molecular Formula C12 H36 O6 Si6
Quantity 250 mg
Synonym Cyclohexasiloxane, 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-, Cyclohexasiloxane, dodecamethyl- (6CI,7CI,8CI,9CI), 2,2,4,4,6,6,8,8,10,10,12,12-Dodecamethylcyclohexasiloxane, Cyclohexadimethylsiloxane, DC 246, DC 246 Fluid, Dodecamethylcyclohexasiloxane, Dodecamethylhexacyclosiloxane, Dow Corning 246, Dow Corning 246 Fluid, Hexadecamethylcyclohexasiloxane, SF 1258, SH 246, SH 246 Fluid, Xiameter PMX 0246
InChI Formula InChI=1S/C12H36O6Si6/c1-19(2)13-20(3,4)15-22(7,8)17-24(11,12)18-23(9,10)16-21(5,6)14-19/h1-12H3
SMILES C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
IUPAC Name 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-1,3,5,7,9,11-hexaoxa-2,4,6,8,10,12-hexasilacyclododecane
Molecular Weight (g/mol) 444.92
Formula Weight 444.1127
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.